Learn More
Eppendorf™ Swing Bucket Rotor for Benchtop Centrifuge
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
A-4-38
A-4-44
A-4-62
A-4-62-MTP
A-4-81
A-4-81-MTP
A-8-17
Includes:
4 Hangers for 2 x 50 mL Falcon-G
4 MTP buckets
4 x 100mL rectangular buckets
4 x 250mL rectangular buckets
4 x 500mL rectangular buckets
4 x 85mL buckets
8 x 15mL buckets
Unit Size:
Each
Description
- Accommodates tubes and plates
- Aerosol-tight caps for safe centrifugation of hazardous samples optionally available
- Rotor life time: max. 40,000 cycles
- Rotor, buckets, caps and adapters are autoclavable
Specifications
Specifications
| Type | Swinging Bucket Rotor |
| Max. RCF | 4,500 x g |
| Max. Speed | 5,000 rpm |
| For Use With (Equipment) | A-4-44 |
| Includes | 4 Hangers for 2 x 50 mL Falcon-G |
| No. of Places | 4 |