Learn More
SVL™ Cone/Screw Thread Adapter
Click to view available options
Unit Size:
Pack of 10
Cone Size:
14/23
24/29
Description
- Features an SVL™ screwthread and Quickfit™ ground glass socket
- Thread accepts SVL™ screw caps and couplings. Cone fits Quickfit™ sockets
- Manufactured from temperature and chemical resistant borosilicate glass
Specifications
Specifications
| For Use With (Equipment) | SVL™ Screw Thread and Quickfit™ Ground Glass Cone |
| Length (Metric) | 85 mm |
| Standard Taper Joint No. | 29/32 |