Learn More
Quickfit™ Claisen Still Head Adapter
Description
Similar to the SH4 still heads but with extra socket to accept dropping funnel or stirrer etc.
Specifications
Specifications
| Cone Size | 19/26 |
| For Use With (Equipment) | Used for thermometer, dropping funnel, stirrer |
| Material | Borosilicate Glass |
| Product Type | Claisen still head Adapter |