missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SV2B Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Probably plays a role in the control of regulated secretion in neural and endocrine cells.
Specifications
Specifications
| Antigen | SV2B |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | Sv2b |
| Gene Accession No. | Q7L1I2, Q8BG39 |
| Gene Alias | A830038F04Rik; AI850768; HsT19680; Kiaa0735; mKIAA0735; SV2B; Sv2bb; synaptic vesicle glycoprotein 2 b; synaptic vesicle glycoprotein 2b; Synaptic vesicle protein 2B; synaptic vesicle protein 2B homolog |
| Gene Symbols | Sv2b |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
