missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SUV420H1 Polyclonal Antibody

Product Code. 15564492
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15564492 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15564492 Supplier Invitrogen™ Supplier No. PA540926

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: NNGFNSGSGS SSTKLKIQLK RDEENRGSYT EGLHENGVCC SDPLSLLESR Sequence homology: Dog: 86%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 91%; Rat: 93%.

Histone methyltransferase that specifically trimethylates 'Lys-20' of histone H4. H4 'Lys-20' trimethylation represents a specific tag for epigenetic transcriptional repression. Mainly functions in pericentric heterochromatin regions, thereby playing a central role in the establishment of constitutive heterochromatin in these regions. KMT5B is targeted to histone H3 via its interaction with RB1 family proteins (RB1, RBL1 and RBL2). Plays a role in myogenesis by regulating the expression of target genes, such as EID3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUV420H1
Applications Immunohistochemistry (Paraffin), Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KMT5B
Gene Accession No. Q4FZB7
Gene Alias [histone H4]-lysine20 N-methyltransferase KMT5B; [histone H4]-N-methyl-L-lysine20 N-methyltransferase KMT5B; AA117471; C630029K18Rik; CGI85; CGI-85; histone-lysine N-methyltransferase KMT5B; histone-lysine N-methyltransferase SUV420H1; Kmt5b; lysine (K)-specific methyltransferase 5B; lysine methyltransferase 5B; lysine N-methyltransferase 5B; Lysine-specific methyltransferase 5B; MGC118906; MGC118909; MGC21161; MGC703; si:dkey-12e7.2; Su(var)4-20 homolog 1; suppressor of variegation 4-20 homolog 1; suppressor of variegation 4-20 homolog 1 (Drosophila); suppressor of variegation 4-20 protein-like 1; SUV420; SUV420H1; suv4-20h1; Unknown (protein for MGC:137299); wu:fb97g06; wu:fi57g03; zgc:103527
Gene Symbols KMT5B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SUV420H1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51111
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.