Learn More
Survivin Antibody (91630) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB886JF635
Product Code. 30170366
  • 595.00 EUR / 0.10mL
Description
Survivin Monoclonal antibody specifically detects Survivin in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Survivin
Monoclonal
Janelia Fluor 635
API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
E. coli-derived recombinant human Survivin, Met1-Asp142, Accession # O15392
RUO
Primary
Human
Liquid
Flow Cytometry (Intracellular Staining), CyTOF
91630
50mM Sodium Borate
Mouse
0.1 mL
Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells
332
Store at 4°C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions