missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Surfactant protein D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11839-1-AP-20UL
This item is not returnable.
View return policy
Description
Surfactant protein D (SP-D) is synthesized and secreted by lung epithelial cells. It belongs to group III of the family of C-type lectins and members of this group has overall structure consisting of multiple globular ′head′ regions linked by triple-helical, collagen-like, strands. This group also includes SP-A and the serum proteins man-binding protein, conglutinin and collectin-43, all of which have been shown to bind to the C1q receptor found on a wide variety of cells. Both SP-D and SP-A have been shown to enhance oxygen radical production by alveolar macrophages.Specifications
| Surfactant protein D | |
| Polyclonal | |
| Unconjugated | |
| SFTPD | |
| COLEC7, Collectin 7, Lung surfactant protein D, PSP D, PSPD, SFTP4, SFTPD, SP D, surfactant protein D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20390, 25350, 6441 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P35247, P35248, P50404 | |
| SFTPD | |
| Surfactant protein D Fusion Protein Ag2461 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction