Learn More
Invitrogen™ SureCast™ Gel Handcast Station
Shop All Thermo Scientific ProductsDescription
Leak-free, confident handcasting of your polyacrylamide gels with the Invitrogen™ SureCast™ Handcast System.
Pour your own polyacrylamide gels easily and confidently with the SureCast Handcast System and SureCast Handcast Reagents. The system features durable glass plates that are securely sealed using a single motion load-and-lock mechanism. The unique tilt feature of the SureCast Handcast Station minimizes spillage and enhances loadability of casting solutions during hand casting. SureCast gels are designed for use in the Mini Gel Tank.
Recommended Storage
Room Temperature
Warranty
Up to 12 months
Specifications
Specifications
| For Use With (Equipment) | SureCast Handcast System |
| Gel Size | Mini |
| Product Line | SureCast |
| Type | SureCast Gel Handcast Station |
| Quantity | 1 station |
| Shipping Condition | Room Temperature |