Description
- Seamless liner for greater comfort
- High flexibility and excellent levels of dexterity
- Very good abrasion and tear resistance
- Allows for contact with food and hot materials up to 100°C
- AQL 1.5
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 3 |
| Certifications/Compliance (Europe) | CE 0493|EN 388:2016 +A1:2018|EN407:2020 |EN ISO 21420:2020|EN ISO 374-1:2016|EN ISO 374-5:2016|Food Type: Rubber|Category III|REACH Compliant |
| Chlorinated | No |
| Applications | Industrial |
| Color | Blue |
| Latex Free | No |
| Material | Latex |
| Product Type | Chemical-resistant Gloves |
| Size | 11 |
| EN388 Standard | 3131A |
| Show More |