Learn More
Memmert™ Support Basket with Perforated Mounting Shelf
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
WTB15
WTB24
WTB35
WTB50
Unit Size:
Each
Description
- Use of test tube racks not possible
- Has grid perforations to take clips for conical flasks
Specifications
Specifications
| For Use With (Equipment) | WTB15 |
| Type | Support Basket With Perforated Mounting Shelf (Stainless Steel) |
| Material | Stainless Steel |