Learn More
Superfos™ SuperLift™ Polypropylene Standard with Centering Lid
Product Code. 10228014
Encompass
Click to view available options
Diameter (Metric) Outer:
223.7 mm
Height (Metric):
12.5 mm
Unit Size:
Each
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 10228014
Brand: Superfos™ 1671
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Manufactured from polypropylene. Superfos™ SuperLift™ Polypropylene Standard with Centering Lid includes tamper evidence while the protective collar safeguards the lid to prevent container ovalisation.
  • Manufactured from polypropylene
  • Includes tamper evidence while the protective collar safeguards the lid and prevents container ovalisation.
  • Very good heat resistance
  • Packaging is suitable for hot filling.
  • Notably, plastic packaging become flexible when subjected to hot filling temperatures and care must be taken in relation to stacking immediately after hot filling.
  • Freezing conditions require special resins available on request.
Specifications
Product Type Standard with Centering Lid
Material Polypropylene
Autoclavable Autoclavable
Certifications/Compliance European Union Commission Regulation No. 10/2011/EC amendments
Diameter (Metric) Outer 223.7 mm
For Use With (Equipment) 3.5 and 5.8L Bucket
Height (Metric) 12.5 mm
Includes Standard Lid with Centering Pieces
Quantity 1 Ea.