Learn More
Sulfur in heavy mineral oil, IP 336, Calibration Set F: 0.5000 % (5000ppm), 1.0000 % (10000ppm), 1.5000 % (15000ppm), 2.0000 % (20000ppm), 3.0000 % (30000ppm), 4.0000 % (40000ppm), 5.0000 % (50000ppm). 100 mL ea., Paragon Scientific™
Click to view available options
Quantity:
Set of 7 = x7 100mLs
Unit Size:
Pack of 7
Description
- Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation.
- Highest level of accreditation guarantee, providing the most credible certified data available worldwide.
- Suitable for but not limited to internationally recognised test methods and protocol: ASTM D2622, ASTM D4294, ASTM D5453, ASTM D7039, IP 336, IP 496, IP 497.
- Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data.
- Supplied in 100 mL volume, tamper-evident glass packaging, providing assurance of sample integrity.
- Manufactured in the United Kingdom.
- 12 month shelf life.
Specifications
Specifications
| Quantity | Set of 7 = x7 100mLs |
Nominal values quoted. Certified Values will vary from batch to batch.