missing translation for 'onlineSavingsMsg'
Learn More

Sulforhodamine B Sodium Salt, TRC

Product Code. 30136583
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136583 5 g 5g
17959994 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136583 Supplier Toronto Research Chemicals Supplier No. TRCS7893005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3520-42-1
Molecular Formula C27 H29 N2 O7 S2 . Na
Molecular Weight (g/mol) 580.65
Synonym Ammonium, [6-(diethylamino)-9-(2,4-disulfophenyl)-3H-xanthen-3-ylidene]diethyl-, hydroxide, inner salt, sodium salt (8CI), C.I. Acid Red 52 (6CI), Xanthylium, 3,6-bis(diethylamino)-9-(2,4-disulfophenyl)-, hydroxide, inner salt, sodium salt, Xanthylium, 3,6-bis(diethylamino)-9-(2,4-disulfophenyl)-, inner salt, sodium salt (9CI), AR 52, Acid Leather Red KB, Acid Red 52, Acid Red B-SF, Acid Red XB, Acid Rhodamine, Acid Rhodamine B, Acid Rose B, Acid Rose Red B, Aizen Food Red No. 106, Amacid Rhodamine B, Amido Rhodamine B, Best Acid Rhodamine FB, Brilliant Acid Rhodamine B, Brilliant Superlan Rhodamine 2B, Brilliant Superlan Rhodamine B, C.I. 45100, Colocid Rhodamine B, Colocid Rhodamine BH, Colocid Rhodamine BN, Conacid Red EB, Covasol Red W 4002, Daiwa IJ Red 207H, Daiwa Red 106WB, Dinacid Rhodamine B, Duasyn Acid Rhodamine B 01, Duramine Rhodamine B, Dyacid Red 4B, Dycosacid Rhodamine B, Erio Acid Red XB, Fenazo Pink XXB, Food Color Red No. 106, Food Red 106, Food Red No. 106, Hispacid Brilliant Pink B, Indacid Rhodamine B, Japan Food Red No. 106, Japan Red 106, Japan Red No. 106, KR 620, Kayacyl Rhodamine FB, Kayaku Acid Rhodamine BH, Kayaku Acid Rhodamine FB, Kemacid Rhodamine B, Kiton Red 620, Kiton Red S, Kiton Rhodamine B, Kyton Red, Libacid Rhodamine LB, Lissamine Rhodamine B, Lissamine Rhodamine B 200, Neolan Red E-XB, Nylosan Rhodamine B, Orient Water Red 27, Pacid Rhodamine B, Phloxine rhodamine, Pontacyl Brilliant Pink, Pro-Jet Red OAM, Rakuto Acid Rhodamine, Ravi Acid Rhodamine B, Red 106, Red no. 106, Rhodamine 200S, Rhodamine Acid, Rhodamine B, Rhodamine B 200, San-Ei Acid Red, Sandolan Rhodamine E-B, Sandolan Rhodamine E-B 400, Sanolin Rhodamine B, Simacid Red 23011, Solar Rhodamine B, Sulfacid Brilliant Pink 3B, Sulfacid Brilliant Pink 3B 1150, Sulforhodamin B, Sulforhodamine B, Triacid Rhodamine B, Vibracolor Red ARE 52, Vicoacid Red 52Y, Xylene Red B
IUPAC Name sodium;4-[3,6-bis(diethylamino)xanthen-10-ium-9-yl]benzene-1,3-disulfonate
SMILES [Na+].CCN(CC)c1ccc2c(c3ccc(cc3S(=O)(=O)[O-])S(=O)(=O)[O-])c4ccc(cc4[o+]c2c1)N(CC)CC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C27H30N2O7S2.Na/c1-5-28(6-2)18-9-12-21-24(15-18)36-25-16-19(29(7-3)8-4)10-13-22(25)27(21)23-14-11-20(37(30,31)32)17-26(23)38(33,34)35;/h9-17H,5-8H2,1-4H3,(H-,30,31,32,33,34,35);/q;+1/p-1
Formula Weight 580.1314
Chemical Name or Material Acid Red 52
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.