missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfitocobalamin Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Sulfitocobalamin Sodium Salt |
| Analyte or Component Names | Sulfitocobalamin Sodium Salt |
| Shipping Condition | Blue Ice |
| Molecular Formula | C62H88CoN13NaO17PS |
| Synonym | Co-(Sulfito-κO)cobinamide Dihydrogen Phosphate (ester) Inner Salt 3'-Ester with (5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole-κN3), Cobalaminsulfonic Acid, Sulfiotcobalamine, Vitamin B12-Sulfonic Acid, Cobinamide, hydroxide sulfite (1:1) (salt), dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole, Cobinamide, sulfite (1:1) (salt), dihydrogen phosphate (ester), 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosylbenzimidazole, Cobinamide, sulfite phosphate, 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosylbenzimidazole, Cobinamide, sulfite (1:1) (salt), dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole |
| InChI Formula | InChI=1S/C62H90N13O14P.Co.H2O3S/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;;1-4(2)3/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;(H2,1,2,3)/q;+3;/p-3 |
| Solution Type | Neat |
| SMILES | -NC(CC1(C)C(CCC(N)=O)C2=N/C1=C(C)\C3=NC(C(C(CC(N)=O)C/4(CCC(NCC(OP(O[C@H]5[C@@H](O)[C@@H](N6C=[N+]([Co+]OS(O[Na])=O)C7=C6C=C(C)C(C)=C7)O[C@@H]5CO)([O-])=O)C)=O)C)[N-]C4=C(C)/C(C(CCC(N)=O)C/8(C)C)=NC8=C/2)(C)C(CC(N)=O)(C)C3CCC(N)=O)=O |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 1432.4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?