Learn More
Thermo Scientific Chemicals Sulfamethazine sodium salt
Product Code. 16667032 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16667032 5 g 5g
16677032 25 g 25g
16687032 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16667032 Supplier Thermo Scientific Chemicals Supplier No. 465600050
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1981-58-4
Molecular Weight (g/mol) 300.31
MDL Number MFCD12352107
Synonym Sulfadimidine sodium salt
Specifications
Type Sulfamethazine Sodium Salt
Physical Form Crystalline Powder
Source Synthetic
Infrared Spectrum Conforms
Loss on Drying ≤2 %
Packaging Glass Bottle
Color White
Quantity 5 g
Molecular Formula C12 H13 N4 Na O2 S
Shipping Condition normal conditions
Solubility Information soluble aq. soln.
Formula Weight 300.31
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Sulfamethazine sodium salt