missing translation for 'onlineSavingsMsg'
Learn More

Sulfacetamide sodium salt hydrate, 99%

Product Code. p-7026431
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470557 25 g 25g
11480557 100 g 100g
11490557 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470557 Supplier Thermo Scientific Alfa Aesar Supplier No. B22426.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6209-17-2 | C8H9N2NaO3S | 236.22 g/mol

Sulfacetamide sodium salt hydrate is an anti-infective agent that is used topically to treat skin infections and orally for urinary tract infections. It adapts to cure trachoma, conjunctivitis, keratitis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfacetamide sodium salt hydrate is an anti-infective agent that is used topically to treat skin infections and orally for urinary tract infections. It adapts to cure trachoma, conjunctivitis, keratitis.

Solubility
Freely soluble in water. Insoluble in chloroform, ether, and benzene.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6209-17-2
Molecular Formula C8H9N2NaO3S
Molecular Weight (g/mol) 236.22
MDL Number MFCD00007885
InChI Key PQMSFAORUFMASU-UHFFFAOYSA-M
Synonym sodium sulfacetamide, sulfacetamide sodium monohydrate, sulfacetamide sodium salt hydrate, sulfacetamide sodic hydrate, unii-4nrt660kjq, sulfacetamide sodium usp, cetamide, sulfair forte, sulfair 10, 4nrt660kjq
PubChem CID 6419954
ChEBI CHEBI:63858
IUPAC Name sodium;acetyl-(4-aminophenyl)sulfonylazanide;hydrate
SMILES CC(=O)N([Na])S(=O)(=O)C1=CC=C(N)C=C1

Specifications

Quantity 25 g
Solubility Information Freely soluble in water. Insoluble in chloroform,ether,and benzene.
Formula Weight 236.23 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Sulfacetamide sodium salt hydrate
compliance-icons
Product Identifier
  • Sulfacetamide, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Acetamide, N-[(4-aminophenyl)sulfonyl]-,monosodium salt, monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.