missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SUGP2 Polyclonal Antibody

Product Code. 15577441
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15577441 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15577441 Supplier Invitrogen™ Supplier No. PA535977

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A. 30 μg HeLa whole cell lysate/extract, B. 30 μg HeLa nuclear lysate/extract. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the arginine/serine-rich family of splicing factors. The encoded protein functions in mRNA processing. Alternatively spliced transcript variants have been described. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUGP2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene SUGP2
Gene Accession No. Q8IX01
Gene Alias Arginine/serine-rich-splicing factor 14; KIAA0365; mKIAA0365; putative splicing factor, arginine/serine-rich 14; serine/arginine-rich splicing factor 14; SFRS14; Splicing factor, arginine/serine-rich 14; Srsf14; SUGP2; SURP and G patch domain containing 2; SURP and G-patch domain containing 2; SURP and G-patch domain-containing protein 2
Gene Symbols SUGP2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human SFRS14. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10147
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.