Learn More
Saint-Gobain Suction Bulb
Product Code. 11714508
Click to view available options
:
Color: Rouge
Product Code. 11714508
Brand: Saint-Gobain 10101410
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Quantity 10 Pack
Type Suction Bulb
For Use With (Application) For sampling
Material Rubber
Color Rouge