missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sucrose stearate

Product Code. p-7043283
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15428369

Brand: Thermo Scientific Chemicals J66845.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25168-73-4 | C30H56O12 | 608.766 g/mol

Sucrose stearate is used as an emulsifier in many beauty applications and cosmetics such as makeup products, cleansing formulas. Emulsifiers aid the blending of water and fatty substances, a common ingredient in many foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucrose stearate is used as an emulsifier in many beauty applications and cosmetics such as makeup products, cleansing formulas. Emulsifiers aid the blending of water and fatty substances, a common ingredient in many foods.

Solubility
Soluble in water and ethanol at 75°C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25168-73-4
Molecular Formula C30H56O12
Molecular Weight (g/mol) 608.766
MDL Number MFCD00152822
InChI Key SZYSLWCAWVWFLT-UTGHZIEOSA-N
Synonym sucrose stearate, sucrose, 1-stearate, unii-58rp7ju52k, sucrose stearate ester, alpha-d-glucopyranoside, 1-o-1-oxooctadecyl-beta-d-fructofuranosyl, 1-o-stearoyl-beta-d-fructofuranosyl alpha-d-glucopyranoside, unii-274kw0o50m component, unii-d0951ov2o1 component, unii-l98x941w2b component, 2s,3s,4s,5r-3,4-dihydroxy-5-hydroxymethyl-2-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy oxolan-2-yl methyl octadecanoate
PubChem CID 9898327
IUPAC Name [(2S,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-2-yl]methyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC1(C(C(C(O1)CO)O)O)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in water and ethanol at 75°C.
Formula Weight 608.75
Physical Form Powder
Chemical Name or Material Sucrose stearate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.