Learn More
Sucrose (Brix) Standard, 45%, Reagecon™
Product Code. 13454036 Shop All Reagecon Products
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
Product Code. 13454036
Brand: Reagecon BS45S