missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Suc-Leu-Leu-Val-Tyr-AMC
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Specifications
Specifications
| Quantity | 5 mg |
| Content And Storage | Protect from light, stored under nitrogen ∣ Powder -80°C 2 years, -20°C 1 year ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen) |
| CAS | 94367-21-2 |
| Solubility | DMSO : ≥ 20 mg/mL (26.18 mM) |
| Product Type | Dye Reagents |
| Molecular Weight (g/mol) | 763.88 g/mol |
| Shipping Condition | Room temperature |
| Target | NO Synthase ∣ MMP ∣ Endogenous Metabolite |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction