Learn More
Hoefer™ SUBHT High Throughput Submarine Gel Unit
Product Code. 12655816
Click to view available options
Type:
Buffer Chamber Assembly
Casting Gate
Colored Loading Strip
High Throughput Submarine Gel Unit
Labeled Running Tray
Lid
Replacement High Voltage Lead
Running Tray
Silicone Gasket
Unit Size:
Each
Product Code. 12655816
Brand: Hoefer™ SUBHT
Includes
  • Buffer chamber assembly
  • Safety lid, high-voltage leads
  • Fluorescent Labeled Running Tray with Casting Gates
  • Four 1mm thick 30-well combs
  • Colored Loading Strips
  • Two Buffer Recirculation Ports
Ideal for high throughput screening applications like single strand polymorphism (SSP) analysis following PCR
  • Fast, error-free loading: load an entire 96-well plate using a multichannel pipetter
  • Simplified Analysis: Fluorescent labels for each sample printed on a UV transparent gel tray allows for instant sample identification (matched to a 96-well plate) during post-run visualization
  • High Throughput: Run up to 120 samples (96-well plate plus 24 control and/or marker samples) in 30 minutes
  • Better Reproducibility: Eliminate variability caused by running buffer pH drift and ionic gradient formation with optional buffer recirculation capability
  • Reduced Buffer Needs: Compact tank design needs less buffer to cover gel which not only reduces buffer consumption but provides greater control of voltage gradients and run times
  • Maximum Flexibility: Fully compatible with a wide range of combs and accessories to meet all your high throughput and standard gel analysis needs
  • This high throughput horizontal gel unit can accommodate up to 120 samples and resolve DNA strands from 50-1000 base pairs in 30 minutes
Specifications
  • Gel Dimensions: 15 x 15 cm
  • Maximum Buffer Volume: 1.2L
  • Maximum Power Settings: 300V, 200mA, 60W
  • Maximum Sample Capacity: 120
  • Maximum Running Temperature: 45°C
  • Indoor Use: 4-40°C
  • Humidity: up to 80%
  • Unit Dimensions: 33.5L x 21.5W x 7 cmH
  • Safety Certifications: EN61010-1, CE
Applications:
Single Strand Polymorphism (SSP) analysis
Specifications
Certifications/Compliance EN61010-1, CE
For Use With (Equipment) SUBHT High Throughput Submarine Gel Unit
Type High Throughput Submarine Gel Unit
Includes Buffer chamber assembly, safety lid with high voltage Leads, running tray with Silicone gasket, 1mm thick, 16-well combs, colored loading strips, Buffer recirculation ports
Sample Size 120
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Volume (Fill) Metric 1.2 L
Dimensions (L x W) 13.1 x 8.46 in. (33.5 x 21.5 cm)
Length (English) 13.1 in.
Length (English) Gel 12.9 in.
Length (Metric) 33.5 cm
Length (Metric) Gel 33 cm
Width (English) 8.46 in.
Width (English) Gel 8.4 in.
Width (Metric) 21.5 cm
Width (Metric) Gel 21.5 cm
Height (Metric) 7 cm
Voltage 300 V
Amperage 200 mA