missing translation for 'onlineSavingsMsg'
Learn More

SU11274, MedChemExpress

Product Code. 16441167
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16441167

Brand: Medchem Express HY12014

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SU11274 is a selective Met inhibitor with IC50 of 10 nM, but has no effects on PGDFRβ, EGFR or Tie2.

SU11274 is a selective Met inhibitor with IC50 of 10 nM, but has no effects on PGDFRβ, EGFR or Tie2.

Specifications

Chemical Name or Material SU11274
CAS 658084-23-2
Color Orange
Purity Grade Notes Research
Molecular Formula C28H30ClN5O4S
Quantity 50 mg
Synonym PKI-SU11274
Solubility Information DMSO : ≥ 100 mg/mL (176.03 mM)
SMILES O=S(C1=CC2=C(C=C1)NC(/C2=C/C3=C(C(C(N4CCN(CC4)C)=O)=C(N3)C)C)=O)(N(C)C5=CC=CC(Cl)=C5)=O
Molecular Weight (g/mol) 568.09
Formula Weight 568.09
Percent Purity 98.19%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.