missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STX16, Mouse anti-Human, Clone: 3D12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00008675-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: MATRRLTDAFLLLRNNSIQNRQLLAEQLADDRMALVSGISLDPEAAIGVTKRPPPKWVDGVDEIQYDVGRIKQKMKELASLHDKHLNRPTLDDSSEEEHSpecifications
| STX16 | |
| Monoclonal | |
| Unconjugated | |
| syntaxin 16 | |
| MGC90328/SYN16/hsyn16 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3D12 | |
| In 1x PBS, pH 7.4 | |
| NM_003763 | |
| STX16 | |
| STX16 (NP_003754.2, 1 a.a. ∼ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 8675 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction