Learn More
Cole-Parmer™ Sample Concentrator Needles for Cole-Parmer™ SC-200 Series Sample Concentrator
Keep your sample concentrator up and running with these Needles.
575.00 EUR - 1185.00 EUR
Product Code | Brand | Material | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Size | Price | Quantity & Availability | ||||
11426578
|
Cole-Parmer™
36620-96 |
Stainless Steel | 76 mm |
575.00 EUR
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
11436578
|
Cole-Parmer™
36620-97 |
Stainless Steel | 127 mm |
630.00 EUR
Pack of 100 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11446578
|
Cole-Parmer™
36620-99 |
PTFE | 76 mm |
885.00 EUR
Pack of 100 |
In Stock
Log in to see stock. |
||||
11456578
|
Cole-Parmer™
36620-98 |
PTFE | 127 mm |
1185.00 EUR
Pack of 100 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||||
Beschreibung
- Maintain your sample concentrator with the following needles.
Spezifikation
Sample Concentrator Needle | |
100 Pk. |