Learn More
Cole-Parmer™ Stuart™ Plastic Cover for Compact Orbital Shakers
Product Code. 12934025
Change view
Click to view available options
For Use With (Equipment):
For SGM-250-030K
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12934025 For SGM-250-030K Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12934025 Supplier Cole-Parmer™ Supplier No. 5170099
Only null left
Add to basket
Add to basket
Parts and accessories to meet your shaking mixer application needs.
Specifications
Product Type Lid
Material Acrylic
Color Clear
For Use With (Equipment) For SGM-250-030K
Packaging Quantity 1 Ea.