Learn More
Eppendorf™ Strips for Fixed Angle Rotor for Benchtop Centrifuge
Product Code. 10690032 Shop All Eppendorf Products
Change view
Click to view available options
Includes:
48 x 0.2mL PCR™ tubes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
10690032 48 x 0.2mL PCR™ tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10690032 Supplier Eppendorf™ Supplier No. 5804735005
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For Centrifuge 5810/5810 R
  • Rotor F-45-48-PCR
  • For Centrifuge 5810/5810 R
  • Holds 6 x 8, 6x 5 PCR tube strips or 48 x 0.2mL individual PCR tubes
TRUSTED_SUSTAINABILITY
Specifications
Type Fixed Angle Rotor
Includes 48 x 0.2mL PCR™ tubes
No. of Places 48