Learn More
Cytiva Strip Holder
Product Code. 10783647 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Ettan IPGphor 3 IEF System 110 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10783647 Ettan IPGphor 3 IEF System 110 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10783647 Supplier Cytiva Supplier No. 80641706
Only null left
Specifications
For Use With (Equipment) Ettan IPGphor 3 IEF System 110 mm
Type Strip holder
Includes 6 x Strip holder; 6 x Strip holder cover; 1 x 60 mL Strip holder cleaning solution
Length (Metric) 11 cm