Learn More
Invitrogen™ Streptococcus agalactiae Monoclonal Antibody (Strep13-509.3), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA5-46098
 View more versions of this product
Product Code. 17870446
  • 424.00 EUR / 100µg
Description
MA1-10875 detects Streptococcus agalactiae from Strep agalactiae samples. MA1-10875 has been successfully used in ELISA applications. The MA1-10875 immunogen is UV-inactivated Strep agalactiae cells isolated from mastitic milk.
Once thought to infect only cows, where it produces mastitis, Streptococcus agalactiae is now known to be able to cause serious disease, bacteremia and meningitis, in immunocompromised individuals and in neonates. Infections in the newborn are the most common and are extremely serious. They are of two types. Early onset of infection, usually occuring within 5 days of birth, is manifested by bacteremia and pneumonia. This infection is contracted vertically, as the baby passes through the birth canal. The other type of infection is a meningitis that occurs between the 10th and 60th days of life. The origin of this infection is not known, but it is not believed to be the mother.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Streptococcus agalactiae
Monoclonal
0.9 mg/mL
PBS with no preservative; pH 7.4
Mouse
Protein G
RUO
Bacteria
Antibody
IgG1
ELISA
Strep13-509.3
Unconjugated
s agalactiae; S. agalactiae
UV-inactivated Strep agalactiae cells isolated from mastitic milk.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions