missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptavidin Antibody (S8C12cc), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-10022
This item is not returnable.
View return policy
Description
Streptavidin Monoclonal specifically detects Streptavidin in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications
| Streptavidin | |
| Monoclonal | |
| Unconjugated | |
| SA V1; SA V2; Streptavidin V1; Streptavidin V2 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant streptavidin. | |
| 0.5 mg | |
| Virology Bacteria and Parasites | |
| No cross-reactivity with egg white avidin. Binds to streptavidin without interfering with the biotin-binding reaction. | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
| CyTOF, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| S8C12 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, CyTOF-ready | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction