Learn More
IBA Lifesciences GmbH Strep-Tactin™ Superflow™ High Capacity Cartridge
Strep-Tactin™ Superflow™ high capacity cartridge is a HPLC/FPLC cartridge to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
124.00 EUR - 345.00 EUR
Specifications
Product Type | High Capacity Cartridge |
---|---|
Column Type | High Capacity Cartridge |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15312026
|
IBA Lifesciences GmbH
2-1238-001 |
1 x 5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15392016
|
IBA Lifesciences GmbH
2-1237-001 |
1 x 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
IBA Lifesciences GmbH Strep-Tactin™ Superflow™ High Capacity Cartridge is a reliable one-step purification of Strep-tag™ fusion proteins with unsurpassed purity and bioacitivity.Specifications
High Capacity Cartridge | |
High Capacity Cartridge |
For Research Use Only.