Learn More
Thermo Scientific Chemicals Straight Wall Zirconium Crucible
Product Code. p-6685028
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
10 mL
15 mL
20 mL
25 mL
35 mL
55 mL
75 mL
100 mL
250 mL
500 mL
1000 mL
Diameter (Metric) Outer:
Outside: 101mm
Outside: 127mm
Outside: 27mm
Outside: 33mm
Outside: 45mm
Outside: 46mm
Outside: 47mm
Outside: 51mm
Outside: 59mm
Outside: 82mm
Height (Metric):
18mm
22mm
29mm
34mm
41mm
45mm
59mm
65mm
89mm
missing translation for 'unitSize'
Each
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15403595
missing translation for 'mfr': Thermo Scientific Chemicals 035909.KT
Request Bulk or Custom Format
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description Straight Wall
Material Zirconium
Capacity (Metric) 10 mL
Diameter (Metric) Outer Outside: 27mm
Height (Metric) 34mm
Product Type Zirconium Crucible