Learn More
Masterflex™ Straight Adapter, Barb x NPT(M), HDPE
Click to view available options
Thread Size:
1/4 in. NPT
1/8 in. NPT
Unit Size:
Pack of 10
Description
- Black, HDPE construction
- Easy to use
- Sterilizable via autoclave or ETO
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | HDPE |
| For Use With | 1/4 in. NPT connection, 3/16 in. ID tubing |
| Packaging Quantity | 10 Pack |
| Product Line | Barb x NPT(M) Connector Fittings |
| Tubing Size | 3/16 in. ID |
| Sterilization Method | Autoclave or Ethylene Oxide |
| Thread Size | 1/4 in. NPT |
| Width (Metric) | 14 mm |
| Color | Black |
| Show More |