Learn More
Cytiva Stop Plug
Product Code. 10722834 Shop All Cytiva Products
Change view
Click to view available options
Connections:
5/16 in. Male
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connections unitSize
10722834 5/16 in. Male Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10722834 Supplier Cytiva Supplier No. 18111250
Only null left
Add to basket
Add to basket
Cytiva Stop Plug, 5/16 in. Male
  • Stop Plug
  • 5/16 in.
  • Male
  • For ÄKTA pure 150 and ÄKTA avant 150
  • Material: Polyetheretherketone (PEEK)
  • Pack of 5
TRUSTED_SUSTAINABILITY
Specifications
Type Connector
Connections 5/16 in. Male
Quantity 5/Pk.
Material PEEK
For Use With (Equipment) ÄKTA Pure 150 and ÄKTA Avant 150