missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Stomatin Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA537857
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
Thought to regulate cation conductance. May regulate ACCN1 and ACCN3 gating.
Specifications
| Stomatin | |
| Polyclonal | |
| Unconjugated | |
| STOM | |
| BND7; EPB7; Epb7.2; EPB72; erythrocyte band 7 integral membrane protein; erythrocyte membrane protein band 7.2 (stomatin); erythrocyte protein band 7.2; erythrocyte protein band 7.2; protein 7.2b; erythrocyte surface protein band 7.2; protein 7.2b; STOM; Stomatin | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 2040 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.485 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P27105 | |
| STOM | |
| Peptide with sequence EKRHTRDSEAQR-C, from the N-terminus of STOM (aa 3-14). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction