Learn More
Sampling Systems™ SteriWare™ Disposable Sample Spoons
BIOPHARMA
Product Code. 15879461 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
3 mL
10 mL
Color:
Blue
Red
White
Length (Metric):
173 mm
185 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Product Code. 15879461
Brand: Sampling Systems™ 8088B10S
Sampling Systems™ SteriWare™ Disposable Sampling Spoons are a range of low cost, single use spoons that allow for quick and efficient surface sampling of powders and granules.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Sterility Sterile
Capacity (Metric) 10 mL
Color Blue
Disposable Single-use
Product Type Spoon
Material Polypropylene
Length (Metric) 185 mm