Learn More
Sampling Systems™ SteriWare™ Disposable Ladle
BIOPHARMA
Product Code. 15869441 Shop All Sampling Systems Products
Click to view available options
Color:
Blue
White
Material:
HDPE
Polypropylene
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 40
Product Code. 15869441
Brand: Sampling Systems™ 8053B50
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Disposable Ladle allows for efficient surface sampling of liquids.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • The one piece moulded design has been specially constructed to go through a standard lid bung
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Material Polypropylene
Capacity (Metric) 50 mL
Disposable Single-use
Sterility Non-sterile
Color Blue
Diameter (Metric) Outer 48 mm