Learn More
Fisherbrand™ Sterile Syringes for Single Use
Product Code. 15859152 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
1 mL
3 mL
5 mL
10 mL
20 mL
60 mL
Connection Type:
Leur Slip
Luer Lock
Unit Size:
Pack of 100
Pack of 50
Product Code. 15859152
Edge
Manual syringe without needle for research purposes only. Suitable for use with syringe filters
  • GAMMA sterilization
  • Sterile
  • Individually wrapped in a paper/plastic blister pack
Warranty
5 Years
Specifications
Diameter (Metric) Inner 9.00 mm
Product Type Luer-Lock
Volume (Metric) 3 mL
Connection Type Luer Lock