Learn More
Merck Stericup™ Quick Release Bottle Top Receiver Flasks
Product Code. 15967996 Shop All Merck Products
Change view
Click to view available options
Volume (Metric):
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15967996 250 mL Pack of 12
15977996 500 mL Pack of 12
15987996 1000 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15967996 Supplier Merck Supplier No. S200B02RE
Only null left
Add to basket
Add to basket
Stericup™ Quick Release Receiver Flask for Bottle Top Filtration includes a click-seal confidence cap that enables a secure grip in wet and dry conditions.
Steritop™ bottle top filter units provide a large filter area for rapid sterile filtration of aqueous fluids into narrow-mouth (33 mm) or wide-mouth (45 mm) bottles.
A small tab inside the funnel secures an optional prefilter in-place during filtration.
For added protection against contamination, the threads for bottle attachment have been slightly recessed in case the funnel accidentally touches a work surface.
TRUSTED_SUSTAINABILITY
Specifications
Outlet Fitting 45 mm
Volume (Metric) 250 mL