Learn More
Synergy Healthcare™ SteriBags™ Heat Seal Closure Pouches
Description
- Conforms to BS 6257
- Double folded bottom for extra integrity
- Opening along width
- For use in autoclaves and gamma sterilizers
Specifications
Specifications
| Description | Sterilization pouch with heat seal closure, code H |
| Autoclavable | Autoclavable |
| Certifications/Compliance | Conforms to BS 6257; Code H |
| Color | White |
| For Use With (Application) | For use in porous load pre-vacuum autoclaves as well as gamma sterilizers |
| Material | Kraft Paper |
| Quantity | 500/Pk. |
| Dimensions (L x W) | 380 x 180 mm |
| Dimensions (L x W x H) | 380 x 180 x 95 mm (L x W x D) |
| Product Type | Sterilization Pouch |