Learn More
asecos™ Steel Safety Canister with Dosing Tap
Product Code. p-200003695 Shop All asecos Products
Change view
Click to view available options
Capacity (Metric):
1 L
2 L
5 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17816566 1 L Each
17886486 2 L Each
17866516 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17816566 Supplier asecos™ Supplier No. 23126
The Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.
  • With fine dosing tap made of stainless steel (1.4301), incl. vent, PTFE seal, integrated flame barrier (removable) and plastic protective ring
  • Incl. multilingual safety sticker to designate the contents
Specifications
Product Type Pouring Can
Capacity (English) 0.3 gal.
Capacity (Metric) 1 L
Color Yellow
For Use With (Application) Safe storage and dosing
Material Steel
Quantity 1
Width (English) 6.46 in.
Width (Metric) 164.00 mm
Height (English) 8.46 in.
Height (Metric) 215.00 mm
Weight (English) 1.65 lb.
Weight (Metric) 0.75 kg