missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Staurosporine, MedChemExpress
Description
Staurosporine is a potent, ATP-competitive and non-selective inhibitor of protein kinases with IC50s of 6 nM, 15 nM, 2 nM, and 3 nM for PKC, PKA, c-Fgr, and Phosphorylase kinase respectively. Staurosporine also inhibits TAOK2 with an IC50 of 3 μM. Staurosporine is an apoptosis inducer.
Specifications
Specifications
| Chemical Name or Material | Staurosporine |
| CAS | 62996-74-1 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C28H26N4O3 |
| Quantity | 10 mg |
| Synonym | Antibiotic AM-2282 STS AM-2282 |
| Solubility Information | DMSO : 62.5 mg/mL (133.97 mM; Need ultrasonic) |
| SMILES | O=C(NC1)C2=C1C3=C(C4=C2C5=C(C=CC=C5)N4[C@H]6C[C@@H](NC)[C@@H](OC)[C@]7(C)O6)N7C8=CC=CC=C83 |
| Molecular Weight (g/mol) | 466.53 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?