missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAU1 Monoclonal Antibody (OTI3C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811475S
This item is not returnable.
View return policy
Description
STAU1 Monoclonal antibody specifically detects STAU1 in Human samples. It is validated for Western BlotSpecifications
| STAU1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P30044 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 25824 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3C5 | |
| Unconjugated | |
| PRDX5 | |
| 5830401L18Rik; AW549911; C85792; Double-stranded RNA-binding protein Staufen homolog 1; PPP1R150; protein phosphatase 1, regulatory subunit 150; RP3-470L14.2; STAU; Stau1; staufen (RNA binding protein) homolog 1 (Drosophila); staufen double-stranded RNA binding protein 1; staufen RNA binding protein homolog 1; staufen, RNA binding protein, homolog 1 | |
| Full length human recombinant protein of human PRDX5 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction