Learn More
- Home
- Products
- Facility Safety and Maintenance
- Labels and Label Makers
- Custom Labels
- Brady™ Static Dissipative Thermal Transfer Printable Labels
Brady™ Static Dissipative Thermal Transfer Printable Labels
Product Code. 11533664
Change view
Click to view available options
Height (Metric):
5.1 mm
6.4 mm
7 mm
9.5 mm
11.1 mm
12.7 mm
Vertical Repeat (Metric):
7.6 mm
8.9 mm
9.5 mm
12.1 mm
13.6 mm
15.2 mm
Width (Metric):
6.4 mm
9.5 mm
12.7 mm
13.8 mm
16.5 mm
17.8 mm
19.1 mm
22.9 mm
25.4 mm
31.8 mm
38.1 mm
50.8 mm
Labels per Row:
1
2
3
4
5
7
12
Unit Size:
1 pack
1 roll
25 product options available for selection
| Product Code. | Height (Metric) | Vertical Repeat (Metric) | Width (Metric) | Labels per Row | unitSize |
|---|---|---|---|---|---|
| 11533664 | 5.1 mm | 7.6 mm | 16.5 mm | 4 | 1 roll |
| 11553664 | 5.1 mm | 7.6 mm | 12.7 mm | 5 | 1 roll |
| 11578434 | 5.1 mm | 7.6 mm | 12.7 mm | 1 | 1 roll |
| 11598434 | 5.1 mm | 7.6 mm | 16.5 mm | 1 | 1 roll |
| 11563654 | 6.4 mm | 8.9 mm | 19.1 mm | 4 | 1 roll |
| 11518444 | 6.4 mm | 8.9 mm | 19.1 mm | 1 | 1 roll |
| 11523674 | 6.4 mm | 8.9 mm | 22.9 mm | 1 | 1 roll |
| 11593664 | 6.4 mm | 8.9 mm | 22.9 mm | 3 | 1 roll |
| 11513674 | 6.4 mm | 8.9 mm | 31.8 mm | 1 | 1 roll |
| 11583654 | 6.4 mm | 8.9 mm | 38.1 mm | 2 | 1 roll |
| 11548444 | 6.4 mm | 8.9 mm | 38.1 mm | 1 | 1 roll |
| 11533674 | 6.4 mm | 8.9 mm | 50.8 mm | 1 | 1 roll |
| 11523664 | 6.4 mm | 9.5 mm | 13.8 mm | 2 | 1 roll |
| 11503674 | 6.4 mm | 9.5 mm | 50.8 mm | 1 | 1 roll |
| 11583664 | 6.4 mm | 8.9 mm | 6.4 mm | 7 | 1 roll |
| 11503664 | 6.4 mm | 9.5 mm | 6.4 mm | 12 | 1 roll |
| 11543664 | 7 mm | 9.5 mm | 12.7 mm | 4 | 1 roll |
| 11573664 | 9.5 mm | 12.1 mm | 17.8 mm | 4 | 1 roll |
| 11573654 | 9.5 mm | 12.1 mm | 25.4 mm | 3 | 1 roll |
| 11528444 | 9.5 mm | 12.1 mm | 25.4 mm | 1 | 1 roll |
| 11563664 | 9.5 mm | 12.1 mm | 9.5 mm | 7 | 1 roll |
| 11513664 | 11.1 mm | 13.6 mm | 12.7 mm | 5 | 1 roll |
| 11593654 | 12.7 mm | 15.2 mm | 25.4 mm | 3 | 1 roll |
| 11538444 | 12.7 mm | 15.2 mm | 25.4 mm | 1 | 1 pack |
| 11588434 | 12.7 mm | 15.2 mm | 12.7 mm | 1 | 1 pack |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Material is UL and CSA approved with its respective ribbon
- Ideal for Circuit Board Labeling
- For smooth surfaces
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | UL Recognized, CSA Accepted, Meets surface resistivity values in the recommended range for dissipative ESD packaging materials as defined by ANSI/ESD S541-2008 (between 10 and 10 ohms), Compliant with Directive 2011/65/EU and 2015/863 |
| Adhesive Type | Acrylic, Permanent |
| Coating | None |
| Color | White |
| Finish | Gloss |
| Format | Die-cut |
| Material | Static-dissipative Polyester |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Product Type | Label |
| Sterility | Non-sterile |
| For Use With (Application) | Circuit Board Labeling, Electrical Labeling, General Identification |
| For Use With (Equipment) | BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500 |
| Horizontal Repeat (Metric) | 17.8 mm |
| Labels per Row | 4 |
| Min. Operating Temperature (Metric) | -40°C |
| Model | THT-14-473-10 |
| No. per Pack | 10000 Label(s) / Roll |
| Quantity | 10,000 Labels |
| Shape | Rectangle |
| Size | 16.51 mm (W) x 5.08 mm (H) |
| Temperature Range (English) | -40°F to 356°F |
| Temperature Range (Metric) | -40°C to +120°C |
| Thickness (English) | 0.0034 in. |
| Thickness (Metric) | 0.09 mm |
| Vertical Repeat (Metric) | 7.6 mm |
| Dimensions (LxW) | 16.51 mm (W) x 5.08 mm (H) |
| Length (English) | 0.2 in. |
| Width (English) | 0.65 in. |
| Width (Metric) | 16.5 mm |
| Width (English) Printable Area | 16.51 mm |
| Width (Metric) Printable Area | 16.5 mm |
| Height (English) | 0.2 in. |
| Height (Metric) | 5.1 mm |
| Height (Metric) Printable Area | 5.1 mm |
| Alternate Part Number | THT-14-473-10 |
| Show More Show Less |