missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Staphylococcus aureus Polyclonal Antibody, Biotin

Product Code. 11508551
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11508551 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11508551 Supplier Invitrogen™ Supplier No. PA173174

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The PA1-73174 antibody reacts with Staphylococcus aureus. PA1-73174 has been successfully used in Immunofluorescence.Immunohistochemistry, ELISA, and Western Blot applications are recommended but have not been tested. The PA1-73174 antibody was raised against soluble and structural antigens of the whole bacterium; ATCC #27660. Short term storage, up to 6 months, is liquid at 4ºC. For longer term storage, aliquot and freeze.

Staphylococcus aureus is a type of bacteria that commonly lives on the skin and in the nasal passages of healthy individuals. However, it can also cause a range of infections, from minor skin infections to more serious conditions such as pneumonia, sepsis, and endocarditis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Staphylococcus aureus
Applications Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Biotin
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias MRSA; s aureus; S. aureus; staphylococcus aureus methicillin-resistant
Host Species Rabbit
Immunogen Staphylococcus aureus whole cells.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.