Learn More
Hach™ Addista Multi-Parameter Standard Solution
Product Code. 10090224
Click to view available options
For Use With (Application):
COD, Ammonium, Chloride, TOC
Nitrite, COD, Phosphate
Unit Size:
Each
Product Code. 10090224
Brand: Hach™ LCA705
Only null left
Add to basket
Add to basket
Make sure that analytical results are always correct. Hach™ Addista Multi-Parameter Standard Solution consists of one combined standard/spiking solution and two round robin test solutions. This system for water quality testing is used with Hach LCK cuvette tests.
With Hach Addista standard solutions, users can easily verify that their analytical approach to water quality testing (sampling, sample preparation, pipetting, photometric analysis) delivers correct results. Addista multiparameter solutions are designed for use with Hach LCK curvette tests.
Each kit comes with one standard solution and two solutions with an unknown concentration for participation in a free round-robin test. After successful participation in this quality assurance program, a certificate confirming the good quality of the analytical results is sent. If results are out of specifications, Hach experts will provide support in identifying and solving the issue.
  • Confidence in the results: Continuous documentation of Analytical Quality Assurance measures
  • Less complexity: Each solutions contains several parameters to reduce the number of single parameter standards in your lab
  • Fewer error sources: Ready-to use solution with Hach LCK cuvettes for less handling to eliminate error sources
Specifications
For Use With (Application) COD, Ammonium, Chloride, TOC
For Use With (Equipment) LCK curvette tests
Includes One standard solution and two solutions with an unknown concentration
Quantity 1 Kit
Type Standard Solutions