Learn More
Tenak™ Standard Rack for Upright Freezer, 32 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
96.55 EUR - 121.00 EUR
Specifications
Material | Stainless Steel |
---|---|
For Use With (Equipment) | Upright Freezers |
Height (Metric) | 32 mm (Box) |
Product Code | Brand | Holds | Array | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Array | Price | Quantity & Availability | ||||
15200644
|
Tenak™
TE23050 |
Boxes (15) | 5 x 3 |
96.55 EUR
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15200654
|
Tenak™
TE23080 |
Boxes (25) | 5 x 5 |
112.00 EUR
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15210644
|
Tenak™
TE23052 |
Boxes (18) | 6 x 3 |
102.00 EUR
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15210654
|
Tenak™
TE23082 |
Boxes (30) | 6 x 5 |
111.00 EUR
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15220644
|
Tenak™
TE23054 |
Boxes (21) | 7 x 3 |
107.00 EUR
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15220654
|
Tenak™
TE23084 |
Boxes (35) | 7 x 5 |
116.00 EUR
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15230644
|
Tenak™
TE23056 |
Boxes (24) | 8 x 3 |
111.00 EUR
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15230654
|
Tenak™
TE23086 |
Boxes (40) | 8 x 5 |
121.00 EUR
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15240644
|
Tenak™
TE23060 |
Boxes (20) | 5 x 4 |
102.00 EUR
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15250644
|
Tenak™
TE23062 |
Boxes (24) | 6 x 4 |
107.00 EUR
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15260644
|
Tenak™
TE23064 |
Boxes (28) | 7 x 4 |
111.00 EUR
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15270644
|
Tenak™
TE23066 |
Boxes (32) | 8 x 4 |
113.00 EUR
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15280644
|
Tenak™
TE23068 |
Boxes (36) | 9 x 4 |
116.00 EUR
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15290644
|
Tenak™
TE23070 |
Boxes (40) | 10 x 4 |
121.00 EUR
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||||
Description
- Measure the inside height and depth of your freezer when choosing the right rack
- Racks can be used with all freezer brands
- Constructed of stainless steel
Specifications
Stainless Steel | |
Upright Freezers | |
32 mm (Box) |