Learn More
Tenak™ Standard Rack for Upright Freezer, 32 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
Brand:  Tenak™ TE23060
Additional Details : Weight : 2.20000kg
 View more versions of this product
Product Code. 15240644
  • 102.00 EUR / Each
Estimated Shipment: 22-07-2024
Description
  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
Specifications
Specifications
5 x 4
Stainless Steel
560 x 139 x 180 mm
Boxes (20)
Upright Freezers
32 mm (Box)