Learn More
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco™ Injectors
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
- Square tip to prevent damage to the injector
- Wide range of volumes
- Precision made from borosilicate glass and stainless steel
- Robust design and easy-to-read markings
Specifications
Specifications
| Description | 250uL |
| Product Type | HPLC Autosampler Syringe |
| Disposable | Reusable |
| Volume (Metric) | 250 μL |
| For Use With (Equipment) | Injector |