Learn More
Kinematica Standard Dispersing Aggregate for PT3100 D
Product Code. 15515369 Shop All Kinematica Products
Change view
Click to view available options
Speed:
8 m/s
Unit Size:
Each
Product Code. Speed unitSize
15515369 8 m/s Each
1 options
Product Code. 15515369
Brand: Kinematica 11030298
Only null left
Add to basket
Add to basket
Uses two rows of teeth, is suitable for a large spectrum of laboratory applications, and is reliable and efficient.
  • Selection and applications:
    • All standard dispersing tasks
    • Grinding of animal and human tissue samples in a variety of fluids and volumes
    • Sample preparation for the extraction and dissolution of organic materials
    • Sample preparation for medical diagnostic procedures as well as DNA analysis
    • Extractions of systems / substances from vegetable-derived samples
    • Disruption of animal and human cells
    • Deagglomeration of united cell structures
    • Intensive mixing
Specifications
Product Type Standard Dispersing Aggregate
Speed 8 m/s
Volume Range (Metric) 0.3 to 10 mL
Diameter (Metric) Probe 7 mm
Length (Metric) 101 mm
Model PT-DA 07 / 2EC-F101
Packaging Quantity 1 Ea.